missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ErbB3/Her3 [p Tyr1289] Antibody (0004), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-30737-20ul

 View more versions of this product

Product Code. 30042700

  • £148.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ErbB3/Her3
Monoclonal
Unconjugated
0.2 μm filtered solution in PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot
0004
Western Blot 1:2000-1:20000
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Tyr1289 of the Human ErbB3/Her3
20 μL
Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
2065
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.