missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£170.00 - £418.00
Specifications
| Antigen | ErbB2/Her2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18213391
|
Novus Biologicals
NB100-81957-0.025ML |
0.025 mL |
£170.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18706573
|
Novus Biologicals
NB100-81957 |
0.1 mL |
£418.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB2/Her2 Polyclonal specifically detects ErbB2/Her2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ErbB2/Her2 | |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| ERBB2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Tumor Suppressors | |
| P04626 | |
| 2064 | |
| IgG | |
| Affinity Purified | |
| 185 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title