missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB2/Her2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81957-0.025ML
This item is not returnable.
View return policy
Description
ErbB2/Her2 Polyclonal specifically detects ErbB2/Her2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ErbB2/Her2 | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian) | |
| Rabbit | |
| 185 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cellular Markers, Tumor Suppressors | |
| 2064 | |
| Human, Rat, Mouse | |
| IgG |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P04626 | |
| ERBB2 | |
| The antiserum was produced against synthesized non-phosphopeptide derived from human HER2 around aa. 1248 (P-E-Y-L-G). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| HER2 (specific to a.a. 1248) detects endogenous levels of total HER2 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction