missing translation for 'onlineSavingsMsg'
Learn More

ErbB2/Her2 Antibody (4D5-8 (trastuzumab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody

£229.00 - £467.00

Specifications

Antigen ErbB2/Her2
Clone 4D5-8 (trastuzumab)
Concentration 1 mg/mL
Applications Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18671767
View Documents
Novus Biologicals
NBP2-75895SS
0.025 mg
£229.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18676707
View Documents
Novus Biologicals
NBP2-75895
0.2 mg
£467.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications

Specifications

ErbB2/Her2
1 mg/mL
Monoclonal
Purified
RUO
Human
biosimilar, biosimilars, CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
A431 cells (human epidermoid carcinoma) (over)expressing EGFR.
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
4D5-8 (trastuzumab)
Flow Cytometry, ELISA, Immunohistochemistry
Unconjugated
Human
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors
PBS
2064
IgG1 κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.