Learn More
ErbB2/Her2 Antibody (4D5-8 (trastuzumab)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP2-75895SS
 View more versions of this product
Product Code. 18671767
  • £229.00 / 0.03mg
Description
ErbB2/Her2 Monoclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ErbB2/Her2
Monoclonal
1 mg/mL
PBS
Human
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, Immunohistochemistry
4D5-8 (trastuzumab)
Unconjugated
biosimilar, biosimilars, CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
A431 cells (human epidermoid carcinoma) (over)expressing EGFR.
0.025 mg
Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Stem Cell Markers, Tumor Suppressors
2064
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG1 κ