missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EquiPhi29™ DNA Polymerase

Product Code. 17360680
Change view
Click to view available options
Quantity:
1000 U
250 U
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17360680 1000 U -
15973292 250 U Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 17360680 Supplier Thermo Scientific™ Supplier No. A39391
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution.

Thermo Scientific EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution. This enzyme is significantly improved over phi29 DNA polymerase in protein thermostability, reaction speed, product yield, and amplification bias, while retaining all the benefits of the wild-type enzyme.

EquiPhi29 DNA Polymerase possesses strong strand displacement activity and 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of EquiPhi29 DNA Polymerase
• Highest processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
• Low amplification bias
• Extremely high yields of amplified DNA, even from small amounts of template
• Highly accurate DNA synthesis in a short time 

Applications
EquiPhi29 DNA Polymerase may be used in a wide variety of applications including:
• Unbiased whole genome amplification (WGA)
• Rolling circle amplification (RCA)
• Protein-primed DNA amplification
• Cell-free cloning of lethal DNA
In situ genotyping with padlock probes
• RNA-primed DNA amplification

Notes
1. The addition of pyrophosphatase to the reaction mixture with EquiPhi29 DNA Polymerase may further enhance DNA synthesis
2. Because of the enzyme's 3'→5' proofreading exonuclease, 3’-modified primers are highly recommended.
3. EquiPhi29 DNA Polymerase can amplify DNA in a wide range of temperatures (30–45°C), with an optimal reaction temperature of 42°C.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stand-alone enzyme
For Use With (Application) MDA-WGA, RCA
Polymerase EquiPhi29 DNA Polymerase
Quantity 1000 U
Content And Storage

• EquiPhi29 DNA Polymerase (100 μL at 10 U/μL)
• 10X EquiPhi29 DNA Polymerase Reaction Buffer (1.0 mL)
• 110 mM DTT (0.25 mL)

Store at –15 to –25°C.

Optimal Reaction Temperature 42°C
Shipping Condition Dry Ice
Concentration 10 U/μL
Reaction Time 2 hours

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.