missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ epT.I.P.S.™ Sets - Pipette Tips

Product Code. 17302910 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
0.25 to 2.5 mL
0.5 to 20 μL
2 to 200 μL
20 to 300 μL
50 to 1000 μL
50 to 1250 μL
Color Code:
Blue
Dark Gray
Green
Light Gray
Medium Gray
Orange
Red
Yellow
Unit Size:
Pack of 240
Pack of 480
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Code Unit Size
17302910 50 to 1000 μL Blue Pack of 480
17352900 0.1 to 10 μL Dark Gray Pack of 480
17362900 0.1 to 20 μL Medium Gray Pack of 480
17372900 0.5 to 20 μL Light Gray Pack of 480
17382900 2 to 200 μL Yellow Pack of 480
17392900 20 to 300 μL Orange Pack of 480
17312910 50 to 1250 μL Green Pack of 480
17322910 0.25 to 2.5 mL Red Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 17302910 Supplier Eppendorf™ Supplier No. 0030076346
  • ONLINE EXCLUSIVE PRICE
    £63.25
    £58.80 / Pack of 480
    Save £4.45
    7% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

Eppendorf epT.I.P.S.™ Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organized pipetting in laboratories.

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • The Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Model epTIPS
Length (Metric) 71 mm
Certifications/Compliance EN ISO 8655, DIN EN ISO 11607-1 and 2 and DIN EN 868-2 to 10.
Clean Claims Eppendorf Quality (Non-sterile)
For Use With (Application) Pipetting liquids, dispensing liquids, mixing liquids, filling plates and reaction vessels, filling gels for electrophoresis, phase extraction and removal of supernatants.
Format Box
Tip Style Standard
Packaging 480 Tips (5 Trays x 96 Tips), 1 Reusable Box
Quantity 480 Pk.
Product Type Pipette Tips
Includes Disposable Racks, Reloads
Color Code Blue
Disposable Single-use
For Use With (Equipment) Air-Cushion System
Graduated Yes
Product Line epT.I.P.S.
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.