missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epsilon 1 Tubulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58223
This item is not returnable.
View return policy
Description
Epsilon 1 Tubulin Polyclonal specifically detects Epsilon 1 Tubulin in Human, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Epsilon 1 Tubulin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dJ142L7.2, Epsilon-tubulin, FLJ22589, FLJ44203, TUBEepsilon-tubulin, tubulin epsilon chain, tubulin, epsilon 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 100%; Rabbit: 92%; Equine: 85%; Xenopus: 78%; Pig: 78%; Chicken: 76%. | |
| Human, Mouse, Rat, Canine, Drosophila, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9UJT0 | |
| TUBE1 | |
| Synthetic peptides corresponding to TUBE1(tubulin, epsilon 1) The peptide sequence was selected from the middle region of TUBE1. Peptide sequence HLHHYLQVEGMEESCFTEAVSSLSALIQEYDQLDATKNMPVQDLPRLSIA. | |
| 100 μL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Tumor Suppressors | |
| 51175 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction