missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPS8L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56615
This item is not returnable.
View return policy
Description
EPS8L1 Polyclonal specifically detects EPS8L1 in Human samples. It is validated for Western Blot.
Specifications
| EPS8L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DRC3EPS8R1, epidermal growth factor receptor kinase substrate 8-like protein 1, Epidermal growth factor receptor pathway substrate 8-related protein 1, EPS8-like 1, EPS8-like protein 1, EPS8-related protein 1, FLJ20258, MGC23164, MGC4642 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54869 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TE68 | |
| EPS8L1 | |
| Synthetic peptides corresponding to EPS8L1(EPS8-like 1) The peptide sequence was selected from the middle region of EPS8L1. Peptide sequence LQKEELRAVSPEEGARVYSQVTVQRSLLEDKEKVSELEAVMEKQKKKVEG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Equine: 100%; Human: 100%; Bovine: 92%; Mouse: 92%; Zebrafish: 92%; Guinea pig: 85%; Rat: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction