Learn More
Eppendorf™ Tubes™ 5.0 mL
Use these tubes, available with screw cap or snap cap, for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ are ideal for working with medium–sized sample volumes. Available in Amber for light-sensitive samples.
Brand: Eppendorf™ 0030122321
63.65 GBP valid until 2024-05-31
Use promo code "21834" to get your promotional price.
Additional Details : Weight : 0.99000kg
Description
Use these tubes for the simple and safe processing of sample volumes up to 5.0mL. Eppendorf Tubes™ represent the perfect option for working with medium–sized sample volumes.
- Conical bottom allows usage in existing adapters and racks.
- Made from transparent polypropylene for easy visibility of pellets.
- Manufactured without use of slip agents, plasticizers and biocides and do not interfere with products.
- Sample evaporation during storage and incubation is minimized by precise lid sealing.
- Accidental lid opening from -86°C to +80°C prevented by hinged lids.
- Centrifugation stability of up to 25,000 x g prevents tube breakage.
- Available in batch-certified Eppendorf, PCR clean, Sterile and Biopur™ purity grades.
- Available in Protein LoBind and DNA/RNA LoBind material for maximum recovery of valuable samples.
- For Cell culture, Isolation of plasmid DNA and total RNA, Liquid handling.
Grade:
- Standard (Eppendorf) Quality = non-sterile
- PCR Clean = non-sterile, Human DNA-free, DNase-free, RNase-free, PCR inhibitor-free,
- Sterile= sterile, Pyrogen-free (endotoxin-free)
- Biopur = sterile, Human DNA-free, DNA-free (human + bacterial DNA), DNase-free, RNase-free, PCR inhibitor-free, ATP-free, Pyrogen-free (endotoxin-free)
- PCR clean and sterile = sterile, Human DNA-free, DNase-free, RNase-free, PCR inhibitor-free, Pyrogen-free (endotoxin-free)
- Forensic DNA Grade = non-sterile, Human DNA-free, DNase-free, RNase-free, PCR inhibitor-free
Specifications
Clear | |
Polypropylene | |
200 Tubes (2 Bags × 100 Tubes) | |
Sterile | |
Pyrogen, Dnase, Rnase, human and bacterial DNA free | |
Screw Cap | |
Yes | |
25000 × g |
5 mL | |
Easy processing of samples up to 5.0 mL, DNA isolation, especially with samples of limited quantity of source material, Preparation of mastermixes and buffers, Cell and tissue lysis | |
Bags | |
0.169 oz. | |
Conical | |
-86°C to +100°C | |
Microcentrifuge Tube | |
Sterile, Pyrogen-, DNase-, RNase-, Human and Bacterial DNA-free |