Learn More
Eppendorf™ Heat Sealer Film and Foils
Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination
Brand: Eppendorf™ 0030127870
Code : 97
Additional Details : Weight : 0.54430kg
Description
- Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
- Transparent heat sealing film allows sample monitoring in the plates
- Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
- Foil stripper tool removes heat-sealed film and foils completely
- Certified free of DNA, DNase, RNase, and PCR inhibitors
- Autoclavable
Specifications
Autoclavable | |
High-strength adhesive that forms tight seals to minimize sample evaporation | |
Yes |
100 Pk. | |
Storage Film |