missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Epoch 2 Microplate Spectrophotometer
GREENER_CHOICE

Product Code. 17247809 Shop All Agilent BioTek Products
Click to view available options
Model:
EPOCH2NS
EPOCH2NSC
EPOCH2TS
EPOCH2TSC
Includes:
Cuvette Port
No Touchscreen, No Cuvette Port
Touchscreen only
Touchscreen; Cuvette port
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17247809

Brand: Agilent BioTek EPOCH2NSCSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. For Research Use Only. Not for use in diagnostic procedures.

The BioTek Epoch 2 microplate spectrophotometer delivers excellent performance for UV-Vis absorbance measurements. Measurements can be performed in 6- to 384-well microplates, cuvets, and in microvolume samples with the available Take3 microvolume plate. The broad wavelength range enables applications from nucleic acid and protein quantification in the low UV to microbial growth assays higher wavelengths

Epoch 2 is easy to use and the optional touch screen interface makes it simple to choose predefined protocols or to define custom programs. Endpoint, kinetic, spectral scanning and well area scanning modes, plus incubation and shaking enable wide-ranging applications.

TRUSTED_SUSTAINABILITY

Specifications

Detector Type Photodiode
Light Source Xenon Flash Lamp
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Accuracy ±2 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Height (English) 22 in.
Hertz 50/60 Hz
Includes Cuvette Port
Model EPOCH2NSC
Wavelength Range 200 to 999 nm, 1 nm increments
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.