Learn More
Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-144-P0
Description
EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.Host Species: Mouse
Clone: VU-1D9
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: Small cell lung carcinoma cells
Molecular Weight: 40-43 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| Epithelial Specific Antigen/Ep-CAM Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| VU-1D9 | |
| Unconjugated | |
| Small cell lung carcinoma cells | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.