missing translation for 'onlineSavingsMsg'
Learn More

Epithelial Specific Antigen/Ep-CAM Ab-1 Mouse Monoclonal Antibody, Epredia™

Product Code. p-4541952 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Unit Size:
100µL
1mL
200µL
500µL
7mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conjugate Form Concentration unitSize
12619836 100 μL Biotin Concentrated 200μg/mL 100µL
12629836 500 μL Biotin Concentrated 200μg/mL 500µL
12609836 1 mL Biotin Concentrated 200μg/mL 1mL
12649836 100 μL Unconjugated Concentrated 200μg/mL 100µL
12659836 100 μL Unconjugated Concentrated 1mg/mL 100µL
12669836 200 μL Unconjugated Concentrated 1mg/mL 200µL
11524584 500 μL Unconjugated Concentrated 200μg/mL 500µL
12639836 1 mL Unconjugated Concentrated 200μg/mL 1mL
12689836 7 mL Unconjugated Prediluted - 7mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12619836 Supplier Epredia Supplier No. MS144B0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epithelial Specific Antigen/Ep-CAM Ab-1, Mouse Monoclonal Antibody.

EGP40 is a 40 kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.

Host Species: Mouse

Clone: VU-1D9

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma cells

Molecular Weight: 40-43 kDa

Positive Control: Breast CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epithelial Specific Antigen/Ep-CAM Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VU-1D9
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Small cell lung carcinoma cells
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.