Learn More
Epidermal Growth Factor Receptor (EGFR) Rabbit Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ EGFR, Rabbit Monoclonal Antibody.
£236.64 - £1505.00
Specifications
| Clone | EP38Y |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Price | Quantity & Availability | ||||
|
12693637
|
Epredia
RM-2111-R7 |
7 mL | Ready-to-Use | ||||||
|
12673637
|
Epredia
RM-2111-RQ |
12 mL | Ready-to-Use | ||||||
|
12603647
|
Epredia
RM-2111-S0 |
100 μL | Supernatant | ||||||
|
12663637
|
Epredia
RM-2111-S |
1 mL | Supernatant | ||||||
Description
EGFR is a type I receptor tyrosine kinase with sequence homology to c-erbB1, B2, B3, B4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-alpha (TGF-alpha), Heparin-binding EGF (HB-EGF), amphiregulin, betacellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, colon, lung, stomach, head and neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.Host Species: Rabbit
Clone: EP38Y
Species Reactivity: Human. Others not tested.
Ig Isotype: Rabbit IgG
Epitope: Not determined
Immunogen: A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR
Molecular Weight: 175 kDa
Positive Control: Squamous carcinoma
Cellular Localization: Cell membrane and occasional cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| EP38Y | |
| Monoclonal | |
| Rabbit | |
| Primary |
| Immunohistochemistry | |
| Unconjugated | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.