missing translation for 'onlineSavingsMsg'
Learn More

Epidermal Growth Factor Receptor (EGFR) Rabbit Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ EGFR, Rabbit Monoclonal Antibody.

Brand:  Epredia RM-2111-S

 View more versions of this product

Product Code. 12663637

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

EGFR is a type I receptor tyrosine kinase with sequence homology to c-erbB1, B2, B3, B4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-alpha (TGF-alpha), Heparin-binding EGF (HB-EGF), amphiregulin, betacellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, colon, lung, stomach, head and neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Rabbit

Clone: EP38Y

Species Reactivity: Human. Others not tested.

Ig Isotype: Rabbit IgG

Epitope: Not determined

Immunogen: A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR

Molecular Weight: 175 kDa

Positive Control: Squamous carcinoma

Cellular Localization: Cell membrane and occasional cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

anti-EGFR
Monoclonal
Unconjugated
A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR
Primary
Supernatant
Immunohistochemistry
EP38Y
Rabbit
1 mL
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.