missing translation for 'onlineSavingsMsg'
Learn More

(-)-Epicatechin Gallate, TRC

Product Code. 30133325
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133325 25 mg 25mg
30012198 1 mg 1mg
30010979 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133325 Supplier Toronto Research Chemicals Supplier No. TRCE58226525MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257-08-5
Molecular Formula C22 H18 O10
Molecular Weight (g/mol) 442.37
Synonym Benzoic acid, 3,4,5-trihydroxy-, 2-(3,4-dihydroxyphenyl)-3,4-dihydro-5,7-dihydroxy-2H-1-benzopyran-3-yl ester, (2R-cis)-, Epicatechol, 3-gallate, (-)- (8CI), Epicatechol, gallate (6CI), Gallic acid, 3-ester with epicatechol, (-)- (8CI), (-)-Epicatechin 3-O-gallate, (-)-Epicatechin 3-gallate, (-)-Epicatechin gallate, (-)-Epicatechol gallate, (-)-epi-Catechin 3-O-gallate, 3-Galloyl-(-)-epicatechin, 3-Galloyl-(2R,3R)-(-)-Epicatechin, 3-O-Galloyl-(-)-epicatechin, 3-O-Galloylepicatechin, ECG, Epicatechin 3-O-gallate, Epicatechin gallate, Epicatechol 3-gallate, L-ECG, L-Epicatechin gallate, epi-Catechin 3-O-gallate
IUPAC Name [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4ccc(O)c(O)c4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H18O10/c23-11-6-14(25)12-8-19(32-22(30)10-4-16(27)20(29)17(28)5-10)21(31-18(12)7-11)9-1-2-13(24)15(26)3-9/h1-7,19,21,23-29H,8H2/t19-,21-/m1/s1
Formula Weight 442.09
Percent Purity >95%
Chemical Name or Material (-)-Epicatechin gallate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.