missing translation for 'onlineSavingsMsg'
Learn More

epi-Lactose, TRC

Product Code. 30137009
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137009 25 mg 25mg
17988324 10 mg 10mg
17909464 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137009 Supplier Toronto Research Chemicals Supplier No. TRCL11410025MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50468-56-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.3
Synonym 4-O-β-D-galactopyranosyl-D-mannose, 4-O-β-D-galactopyranosyl-mannose, Epilactose;
IUPAC Name (2S,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal
SMILES OC[C@H]1O[C@@H](O[C@H]2C(O)[C@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H22O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h1,4-12,14-21H,2-3H2/t4-,5-,6-,7-,8+,9+,10-,11-,12+/m1/s1
Formula Weight 342.12
Percent Purity >95%
Chemical Name or Material Epilactose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.