missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB6 Rabbit anti-Human, Clone: 31, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | EphB6 |
|---|---|
| Clone | 31 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210007
|
Novus Biologicals
NBP2-89327-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243967
|
Novus Biologicals
NBP2-89327-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| EphB6 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911 | |
| Recombinant Human EphB6 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 31 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| 0.2 um filtered solution in PBS | |
| 2051 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title