missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphB6 Rabbit anti-Human, Clone: 31, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89327-100UL
This item is not returnable.
View return policy
Description
EphB6 Monoclonal specifically detects EphB6 in Human samples. It is validated for Sandwich ELISA Detection.
Specifications
| EphB6 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 31 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911 | |
| Recombinant Human EphB6 Protein | |
| 100 μg | |
| Protein Kinase | |
| 2051 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction