Learn More
Invitrogen™ EPHA1 Recombinant Human Protein
EPHA1 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family.
Brand: Invitrogen™ PV4093
Code : NEW
Additional Details : Weight : 0.00900kg
Description
EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system.
Specifications
NP_005223.2 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
EPHA1, EPH | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EPH Receptor Kinase Family | |
Recombinant | |
Baculovirus |