missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EPHA1 Recombinant Human Protein
EPHA1 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family.
£1602.00 - £6900.00
Specifications
Accession Number | NP_005223.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 74.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448603
|
Invitrogen™
PR7498A |
100μg |
£1602.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10518303
|
Invitrogen™
PV4093 |
1mg |
£6900.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system.
Specifications
NP_005223.2 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
EPH Receptor Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.4kDa | |
RUO - research use only | |
EPHA1, EPH | |
Human | |
GST Tag | |
Frozen |