missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epcoritamab Chimeric Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
£483.00 - £2005.00
Specifications
| Antigen | Epcoritamab Chimeric |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30283405
![]() |
Invitrogen™
MA560133 |
100 μg |
£483.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283128
|
Invitrogen™
MA560134 |
1 mg |
£2005.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Epcoritamab (DuoBody™-CD3xCD20) is a proprietary bispecific antibody created using Genmab's DuoBody technology. It targets CD20 on the B-cells, a clinically well-validated target that is expressed in a wide variety of B-cell malignancies. Epcoritamab is currently under investigation in a Phase I/II study for multiple hematological B cell malignancies.Specifications
| Epcoritamab Chimeric | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| GEN3013 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title