missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epcoritamab Chimeric Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560134
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Epcoritamab (DuoBody™-CD3xCD20) is a proprietary bispecific antibody created using Genmab's DuoBody technology. It targets CD20 on the B-cells, a clinically well-validated target that is expressed in a wide variety of B-cell malignancies. Epcoritamab is currently under investigation in a Phase I/II study for multiple hematological B cell malignancies.
Specifications
| Epcoritamab Chimeric | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GEN3013 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction