missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax™ Racks - Self-Sealing Pipette Tips
GREENER_CHOICE

Product Code. 16482275 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL S
0.5 to 20 μL
2 to 100 μL
2 to 200 μL
20 to 300 μL
50 to 1000 μL
Color Code:
Blue
Dark Gray
Light Gray
Orange
Yellow
Length (Metric):
34 mm
46 mm
53 mm
55 mm
76 mm
Unit Size:
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Code Length (Metric) unitSize
16482275 20 to 300 μL Orange 55 mm Pack of 960
16442275 0.1 to 10 μL S Dark Gray 34 mm Pack of 960
16452275 0.5 to 20 μL Light Gray 46 mm Pack of 960
16462275 2 to 100 μL Yellow 53 mm Pack of 960
16472275 2 to 200 μL Yellow 55 mm Pack of 960
16492275 50 to 1000 μL Blue 76 mm Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16482275 Supplier Eppendorf™ Supplier No. 0030078730
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eppendorf ep Dualfilter T.I.P.S.™ SealMax Racks provide advanced self-sealing pipette tips in 'PCR clean and Sterile' grade, featuring unique two-phase filters for unparalleled protection against contamination in sensitive lab applications.

  • Reliable protection from liquids, aerosols (1) and biomolecules
  • Certificate of filter efficiency of class ISO 25 E in accordance with DIN EN ISO 29463-5 available
  • No PCR inhibiting additives
  • Protection of your pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20% lower polypropylene consumption per rack
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
  • (1) An aerosol is a colloid of fine solid particles or liquid droplets, in air or another gas.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 to 300 μL
Filtered Filtered
Sterility Sterile
Color Colorless
Autoclavable Not Autoclavable
Length (English) 2.2 in.
Length (Metric) 55 mm
Clean Claims PCR Clean and Sterile
Features PCR Clean
Format Rack
Packaging 960 Tips (10 Racks x 96 Tips)
Quantity 960 Tips
Product Type Self-sealing Pipette Tips
Includes 10 Racks x 96 Tips
Color Code Orange
For Use With (Equipment) Air-Cushion System
Product Line ep DualFilter T.I.P.S
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.