missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ 3C-like Protease (3CLpro) & Papain-like Protease (PLpro) Assay Kits

Product Code. p-9482004
Click to view available options
No. of Reactions:
100 Assays
5 x 100 Assays
Assay:
3C-like Protease (3CLpro) SARS-CoV-2
Papain-like Protease (PLpro)
Papain-like Protease (PLpro) SARS-CoV-2
Unit Size:
Pack of 100
Pack of 500
This item is not returnable. View return policy

Product Code. 17747483

Brand: Invitrogen™ E33970

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Quantify and screen inhibitors of SARS-CoV-2 3C-like protease (3CLpro) and Papain-like Protease (PLpro) with the EnzChek Protease assay kits. Includes all reagents for the determination of protease activity, including a positive control inhibitor.

Easily and quickly quantify inhibitors of SARS-CoV-2 3C-like protease (3CLpro) and papain-like protease (PLpro) using the EnzChek 3CLpro and PLpro Assay kits. These kits can be used to screen and profile inhibitors of 3CLpro and PLpro and include all reagents for the determination of protease activity, including a positive control inhibitor. 3CLpro and PLpro activity can be directly correlated using a 96-well plate and microplate reader.

The EnzChek 3C-like Protease (3CLpro) and Papain-like Protease (PLpro) Assay kits are quick, easy, and direct assays for detecting and quantifying SARS-CoV-2 3-chymotrypsin-like protease (3CLpro) and SARS-CoV-2 papain-like protease (PLPro) activity. These kits can be used to screen and profile inhibitors of 3CLpro and PLpro and include all reagents for the determination of protease activity, including a positive control inhibitor. 3CLpro and PLpro activity can be directly correlated using a 96-well plate and microplate reader.

EnzChek 3CLpro and PLpro Assay kit features include:

Sensitive — detect as little as 3 ng of 3CLpro or 6 ng of PLpro
Quick — less than two hours for results
Simple — easy to use and ideal for high-throughput screening applications

3CLpro, also known as main protease (Mpro), and PLpro are crucial enzymes for processing viral polyproteins, and due to their role in the SARS-CoV-2 virus life cycle have been identified as a promising drug target for blocking viral replication. The homogeneous EnzChek 3CLpro and PLpro Assay kits are ideal for easily screening and quantifying inhibitors of 3CLpro and PLpro.

The EnzChek 3C-like 3CLpro and PLpro Assay kits contain a fluorogenic protease-specific substrate, SARS-CoV-2 3CLpro or PLpro, positive control inhibitor, and assay buffer. Simply add your inhibitor of choice. Protease-catalyzed hydrolysis releases the fluorescently labeled peptide, and protease activity can be directly quantified from the change in fluorescence signal using a microplate reader (ex: 360 nm, em: 460 nm).
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorometric
Product Type Assay
Conjugate DABCYL/EDANS
Kit Contents 3C-like Protease (3CLpro) SARS-CoV-2, Protease Assay Buffer, 3C-like Protease (3CLpro) Fluorescent Substrate, GC376 (positive control inhibitor), Buffer Additive A
Assay 3C-like Protease (3CLpro) SARS-CoV-2
Druggable Target 3C-like Protease (3CLpro)
Excitation/Emission 360/460
No. of Reactions 100 Assays
For Use With (Equipment) Fluorescent Microplate Reader
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.