missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EnzChek™ 3C-like Protease (3CLpro) & Papain-like Protease (PLpro) Assay Kits
Click to view available options
No. of Reactions:
100 Assays
5 x 100 Assays
Assay:
3C-like Protease (3CLpro) SARS-CoV-2
Papain-like Protease (PLpro)
Papain-like Protease (PLpro) SARS-CoV-2
Packungsgröße:
Pack of 100
Pack of 500
Beschreibung
Easily and quickly quantify inhibitors of SARS-CoV-2 3C-like protease (3CLpro) and papain-like protease (PLpro) using the EnzChek 3CLpro and PLpro Assay kits. These kits can be used to screen and profile inhibitors of 3CLpro and PLpro and include all reagents for the determination of protease activity, including a positive control inhibitor. 3CLpro and PLpro activity can be directly correlated using a 96-well plate and microplate reader.
The EnzChek 3C-like Protease (3CLpro) and Papain-like Protease (PLpro) Assay kits are quick, easy, and direct assays for detecting and quantifying SARS-CoV-2 3-chymotrypsin-like protease (3CLpro) and SARS-CoV-2 papain-like protease (PLPro) activity. These kits can be used to screen and profile inhibitors of 3CLpro and PLpro and include all reagents for the determination of protease activity, including a positive control inhibitor. 3CLpro and PLpro activity can be directly correlated using a 96-well plate and microplate reader.
EnzChek 3CLpro and PLpro Assay kit features include:
• Sensitive — detect as little as 3 ng of 3CLpro or 6 ng of PLpro
• Quick — less than two hours for results
• Simple — easy to use and ideal for high-throughput screening applications
3CLpro, also known as main protease (Mpro), and PLpro are crucial enzymes for processing viral polyproteins, and due to their role in the SARS-CoV-2 virus life cycle have been identified as a promising drug target for blocking viral replication. The homogeneous EnzChek 3CLpro and PLpro Assay kits are ideal for easily screening and quantifying inhibitors of 3CLpro and PLpro.
The EnzChek 3C-like 3CLpro and PLpro Assay kits contain a fluorogenic protease-specific substrate, SARS-CoV-2 3CLpro or PLpro, positive control inhibitor, and assay buffer. Simply add your inhibitor of choice. Protease-catalyzed hydrolysis releases the fluorescently labeled peptide, and protease activity can be directly quantified from the change in fluorescence signal using a microplate reader (ex: 360 nm, em: 460 nm).
EnzChek 3CLpro and PLpro Assay kit features include:
• Sensitive — detect as little as 3 ng of 3CLpro or 6 ng of PLpro
• Quick — less than two hours for results
• Simple — easy to use and ideal for high-throughput screening applications
3CLpro, also known as main protease (Mpro), and PLpro are crucial enzymes for processing viral polyproteins, and due to their role in the SARS-CoV-2 virus life cycle have been identified as a promising drug target for blocking viral replication. The homogeneous EnzChek 3CLpro and PLpro Assay kits are ideal for easily screening and quantifying inhibitors of 3CLpro and PLpro.
The EnzChek 3C-like 3CLpro and PLpro Assay kits contain a fluorogenic protease-specific substrate, SARS-CoV-2 3CLpro or PLpro, positive control inhibitor, and assay buffer. Simply add your inhibitor of choice. Protease-catalyzed hydrolysis releases the fluorescently labeled peptide, and protease activity can be directly quantified from the change in fluorescence signal using a microplate reader (ex: 360 nm, em: 460 nm).
Spezifikation
Spezifikation
| Detection Method | Fluorometric |
| Product Type | Assay |
| Conjugate | AMC |
| Kit Contents | Papain-like Protease (PLpro) SARS-Cov-2, Protease Assay Buffer, Papain-like Protease (PLpro) Fluorescent Substrate, GRL0617 (positive control inhibitor), Buffer Additive A |
| Assay | Papain-like Protease (PLpro) |
| Druggable Target | Papain-like Protease (PLpro) |
| Excitation/Emission | 360/460 |
| No. of Reactions | 5 x 100 Assays |
| For Use With (Equipment) | Fluorescent Microplate Reader |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur