missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Enterobacteria Enrichment Broth-Mossel

Product Code. 12693458
Click to view available options
Fill Weight:
10.0 to 10.3 g
100 – 101.5 g
100-101.5 g
Format:
100 mL sirop bottle
125 mL sirop bottle
Universal Bottle
Quantity:
10 x 100 mL
10 x 90 mL
24/Pk.
Unit Size:
Each
Pack of 10
Pack of 24
This item is not returnable. View return policy

Product Code. 12693458

Brand: Thermo Scientific™ BO1156J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A selective enrichment broth for the detection of bile tolerent Gram-negative bacteria in microbiological examination of pharmaceutical products complying with EP/USP/JP/BP specification.

Bottled media for Enterobacteria enrichment media complying with EP/USP/JP/BP specification.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® ARTG
Certifications/Compliance EP/USP/JP/BP
Content And Storage 2°C to 8°C
Country of Origin UK
Format 100 mL sirop bottle
Media Type Prepared Media
Product Type Microbiology Media
Fill Weight 100 – 101.5 g
For Use With (Application) Enrichment Broth for Enterobacteria
Packaging Type 10 sirop bottles bottles with screw cap closures in a box
Quantity 10 x 90 mL
Species Enterobacteria
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.