missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Enrofloxacin Monoclonal Antibody (7F21G1F7E8#1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£367.00 - £916.00

Specifications

Antigen Enrofloxacin
Clone 7F21G1F7E8#1
Concentration 1.6 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17890156
View Documents
Invitrogen™
MA5-46045
100 μg
£367.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11573393
GREENER_CHOICE
View Documents
Invitrogen™
MA110782
500 μg
£467.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17860346
View Documents
Invitrogen™
MA5-46143
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10782 detects Enrofloxacin and its main metabolite, ciproflaxin. MA1-10782 has been successfully used in ELISA applications. The MA1-10782 immunogen is Enrofloxacin conjugated to KLH.

Enrofloxacin is a fluoroquinolone that inhibits the activity of bacterial DNA gyrase. It was the first fluoroquinolone antimicrobial to be used in veterinary medicine and is indicated in therapy of infections by E. coli, Salmonella, Pasteurella, Mycoplasma and Hemophilus species. In the USA, approved use is currently limited to dogs, but approval for use in foodanimals is under consideration.
Specifications

Specifications

Enrofloxacin
1.6 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Enrofloxacin conjugated to KLH.
Primary
-20°C, Avoid Freeze/Thaw Cycles
7F21G1F7E8#1
ELISA
Unconjugated
Mouse
Chemical
Baytril
IgM
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.