Learn More
Invitrogen™ Enrofloxacin Monoclonal Antibody (7F21G1F7E8#1), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46045
 View more versions of this product
Product Code. 17890156
  • £367.00 / 100µg
Description
MA1-10782 detects Enrofloxacin and its main metabolite, ciproflaxin. MA1-10782 has been successfully used in ELISA applications. The MA1-10782 immunogen is Enrofloxacin conjugated to KLH.
Enrofloxacin is a fluoroquinolone that inhibits the activity of bacterial DNA gyrase. It was the first fluoroquinolone antimicrobial to be used in veterinary medicine and is indicated in therapy of infections by E. coli, Salmonella, Pasteurella, Mycoplasma and Hemophilus species. In the USA, approved use is currently limited to dogs, but approval for use in foodanimals is under consideration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Enrofloxacin
Monoclonal
1.6 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein A
RUO
Chemical
Antibody
IgM
ELISA
7F21G1F7E8#1
Unconjugated
Baytril
Enrofloxacin conjugated to KLH.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical