missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ENPP-2/Autotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | ENPP-2/Autotaxin |
|---|---|
| Dilution | Western Blot : 5-10 μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067234
|
Novus Biologicals™
NBP3-30863-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067182
|
Novus Biologicals™
NBP3-30863-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ENPP-2/Autotaxin Polyclonal antibody specifically detects ENPP-2/Autotaxin in Human samples. It is validated for ELISA,Western BlotSpecifications
| ENPP-2/Autotaxin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| Phosphate buffered saline | |
| 5168 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-10 μg/mL, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, Extracellular lysophospholipase D, LysoPLD, PD-IALPHA, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D | |
| Synthetic peptide derived from the ENPP-2/Autotaxin | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title