missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ENPP-2/Autotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30863-100ug
This item is not returnable.
View return policy
Description
ENPP-2/Autotaxin Polyclonal antibody specifically detects ENPP-2/Autotaxin in Human samples. It is validated for ELISA,Western Blot
Specifications
| ENPP-2/Autotaxin | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, ELISA | |
| ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, Extracellular lysophospholipase D, LysoPLD, PD-IALPHA, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D | |
| Synthetic peptide derived from the ENPP-2/Autotaxin | |
| 100 μg | |
| Cardiovascular Biology | |
| 5168 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction