missing translation for 'onlineSavingsMsg'
Learn More

Enoxacin, Thermo Scientific™

A quinolone antibacteral agent which acts on DNA gyrase

£21.65 - £216.00

Chemical Identifiers

CAS 74011-58-8
Molecular Formula C15H17FN4O3
Molecular Weight (g/mol) 320.324
MDL Number MFCD00133308
InChI Key IDYZIJYBMGIQMJ-UHFFFAOYSA-N
Synonym Penetrex™ The Clorox Company; Bactidan
PubChem CID 3229
ChEBI CHEBI:157175
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444399
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J61912.03
1g
ONLINE EXCLUSIVE PRICE
£23.50
£21.65 / 1g
Save £1.85
7% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15454399
SDS View Documents
Thermo Scientific Alfa Aesar
J61912.06
5g
£58.80
5g
View alternative products
Save Up to 
»
Estimated Shipment: 09-09-2026
Add to basket
 
 Restricted Item
15464399
SDS View Documents
Thermo Scientific Alfa Aesar
J61912.14
25g
£216.00
25g
View alternative products
Save Up to 
»
Estimated Shipment: 25-09-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections. It is a bactericidal and its mode of action depends on blocking of bacterial DNA replication by binding itself to an enzyme called DNA gyrase.

Solubility
50 mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide, dimethyl sulfoxide, chloroform and methanol. Insoluble in water.

Notes
Store in cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

74011-58-8
320.324
IDYZIJYBMGIQMJ-UHFFFAOYSA-N
3229
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
C15H17FN4O3
MFCD00133308
Penetrex™ The Clorox Company; Bactidan
CHEBI:157175
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O

Specifications

74011-58-8
C15H17FN4O3
14,3587
50mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide,dimethyl sulfoxide,chloroform and methanol; Insoluble in water.
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O
320.324
CHEBI:157175
Enoxacin
White to Yellow
MFCD00133308
Penetrex™ The Clorox Company; Bactidan
IDYZIJYBMGIQMJ-UHFFFAOYSA-N
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
3229
320.32
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : QN2800000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.