missing translation for 'onlineSavingsMsg'
Learn More

Enoxacin, Thermo Scientific™

A quinolone antibacteral agent which acts on DNA gyrase

Brand:  Thermo Scientific Alfa Aesar J61912.06

Quantity 5g

 View more versions of this product

Product Code. 15454399

  • £58.80 / 5g
 Restricted Item
Estimated Shipment: 10-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enoxacin is an antibacterial agent used in the treatment of gastro enteritis including infectious diarrhea, respiratory tract infections, gonorrhea and urinary tract infections. It is a bactericidal and its mode of action depends on blocking of bacterial DNA replication by binding itself to an enzyme called DNA gyrase.

Solubility
50 mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide, dimethyl sulfoxide, chloroform and methanol. Insoluble in water.

Notes
Store in cool place. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

74011-58-8
320.324
IDYZIJYBMGIQMJ-UHFFFAOYSA-N
3229
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
C15H17FN4O3
MFCD00133308
Penetrex™ The Clorox Company; Bactidan
CHEBI:157175
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O

Specifications

74011-58-8
5g
MFCD00133308
Penetrex™ The Clorox Company; Bactidan
IDYZIJYBMGIQMJ-UHFFFAOYSA-N
1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid
3229
320.32
White to Yellow
C15H17FN4O3
14,3587
50mg/ml in 1 M NaOHSlightly soluble in sodium hydroxide,dimethyl sulfoxide,chloroform and methanol; Insoluble in water.
CCN1C=C(C(=O)C2=CC(=C(N=C21)N3CCNCC3)F)C(=O)O
320.324
CHEBI:157175
Enoxacin
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.