missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enolase 2/Neuron-specific Enolase Antibody (SPM347), Novus Biologicals™
Mouse Monoclonal Antibody
£232.00 - £428.00
Specifications
| Antigen | Enolase 2/Neuron-specific Enolase |
|---|---|
| Clone | SPM347 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18248550
|
Novus Biologicals
NBP2-53158-20UG |
20 μg |
£232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250500
|
Novus Biologicals
NBP2-53158-100UG |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Enolase 2/Neuron-specific Enolase | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2026 | |
| A synthetic peptide corresponding to aa416-433 of human NSE gamma (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| SPM347 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| ENO2 | |
| IgG2b | |
| Protein A or G purified | |
| 50 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts