missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enolase 2/Neuron-specific Enolase Antibody (SPM347), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53158-100UG
This item is not returnable.
View return policy
Description
Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Enolase 2/Neuron-specific Enolase | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| Mouse | |
| 50 kDa | |
| 100 μg | |
| Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 2026 | |
| Store at 4C. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM347 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| ENO2 | |
| A synthetic peptide corresponding to aa416-433 of human NSE gamma (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction