missing translation for 'onlineSavingsMsg'
Learn More

Engeletin, MedChemExpress

Product Code. 16489076
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489076 100 mg 100mg
16461408 5 mg 5mg
16454837 50 mg 50mg
16453866 25 mg 25mg
16418988 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16489076 Supplier Medchem Express Supplier No. HYN0436
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Engeletin is a flavanonol glycoside isolated from hymenaea martiana, inhibits NF-κB signaling-pathway activation, and possesses anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects.

Engeletin is a flavanonol glycoside isolated from hymenaea martiana, inhibits NF-κB signaling-pathway activation, and possesses anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects.

Specifications

Chemical Name or Material Engeletin
CAS 572-31-6
Color White
Purity Grade Notes Research
Molecular Formula C21H22O10
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (230.21 mM)
SMILES OC1=CC(O)=C(C([C@H](O[C@@]2([H])[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O2)[C@@H](C3=CC=C(O)C=C3)O4)=O)C4=C1
Molecular Weight (g/mol) 434.39
Formula Weight 434.39
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.