missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin 1 Mouse anti-Human, Unconjugated, Clone: 2C5B3, Proteintech
Mouse Monoclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | Endothelin 1 |
|---|---|
| Clone | 2C5B3 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
The Endothelin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Endothelin 1. This antibody recognizes human antigen. The Endothelin 1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| Endothelin 1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 1906 | |
| Primary |
| 2C5B3 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title