Learn More
Endothelin 1 Mouse anti-Human, Unconjugated, Clone: 2C5B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67008-1-IG-20UL
Description
The Endothelin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Endothelin 1. This antibody recognizes human antigen. The Endothelin 1 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| Endothelin 1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Flow Cytometry, ELISA | |
| 2C5B3 | |
| Unconjugated | |
| 20 μL | |
| 1906 | |
| IgG1 |