missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endothelin-1 Antibody (TR.ET.48.5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
£169.00
Specifications
| Antigen | Endothelin-1 |
|---|---|
| Clone | TR.ET.48.5 |
| Concentration | 2.5 mg/mL |
| Dilution | Western Blot 1:100 - 1:1000, Flow Cytometry 0.5 μg, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen 1:250, Radioimmunoassay 1:25000 |
| Applications | Western Blot |
Description
Endothelin-1 Monoclonal specifically detects Endothelin-1 in Human, Mouse, Rat, Porcine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay.Specifications
| Endothelin-1 | |
| 2.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1 | |
| EDN1 | |
| IgG1 | |
| Affinity Purified | |
| Not yet tested in other species. |
| TR.ET.48.5 | |
| Western Blot 1:100 - 1:1000, Flow Cytometry 0.5 μg, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen 1:250, Radioimmunoassay 1:25000 | |
| Monoclonal | |
| Mouse | |
| Cell Cycle and Replication | |
| P05305 | |
| 1906 | |
| Pure endothelin-1 conjugated to KLH. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title