missing translation for 'onlineSavingsMsg'
Learn More

Endothelin-1 Antibody (TR.ET.48.5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NB300-526-0.025ml

 View more versions of this product

Product Code. 18531413

  • £169.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Endothelin-1 Monoclonal specifically detects Endothelin-1 in Human, Mouse, Rat, Porcine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endothelin-1
Monoclonal
2.5 mg/mL
Western Blot 1:100 - 1:1000, Flow Cytometry 0.5 μg, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen 1:250, Radioimmunoassay 1:25000
endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1
Mouse
Affinity Purified
RUO
Primary
Not yet tested in other species.
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
TR.ET.48.5
Unconjugated
P05305
EDN1
Pure endothelin-1 conjugated to KLH.
0.025 mL
Cell Cycle and Replication
1906
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only