missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endophilin B1/Bif-1 Antibody (30A882.1.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 26 publications
£168.00 - £391.00
Specifications
| Antigen | Endophilin B1/Bif-1 |
|---|---|
| Clone | 30A882.1.1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
18076805
|
Novus Biologicals
NBP2-24733SS |
0.025 mg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799183
|
Novus Biologicals
NBP2-24733 |
£391.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Endophilin B1/Bif-1 Monoclonal specifically detects Endophilin B1/Bif-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Spécification
| Endophilin B1/Bif-1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Mitochondrial Mediated Pathway | |
| Q9Y371 | |
| 51100 | |
| A full-length human Bif-1 recombinant protein was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 30A882.1.1 | |
| Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Bax-interacting factor 1, Bif-1dJ612B15.2, CGI-61, endophilin-B1, KIAA0491endophilin B1, SH3 domain-containing GRB2-like protein B1, SH3-containing protein SH3GLB1, SH3-domain GRB2-like endophilin B1, SH3-domain, GRB2-like, endophilin B1 | |
| SH3GLB1 | |
| IgG1 κ | |
| Protein G purified | |
| This antibody is reactive to Human. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit