missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endophilin B1/Bif-1 Antibody (30A882.1.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 26 publications
Brand: Novus Biologicals NBP2-24733SS
This item is not returnable.
View return policy
Description
Endophilin B1/Bif-1 Monoclonal specifically detects Endophilin B1/Bif-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Endophilin B1/Bif-1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Simple Western 1:200, Immunohistochemistry 5ug/ml, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Bax-interacting factor 1, Bif-1dJ612B15.2, CGI-61, endophilin-B1, KIAA0491endophilin B1, SH3 domain-containing GRB2-like protein B1, SH3-containing protein SH3GLB1, SH3-domain GRB2-like endophilin B1, SH3-domain, GRB2-like, endophilin B1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 30A882.1.1 | |
| Unconjugated | |
| Q9Y371 | |
| SH3GLB1 | |
| A full-length human Bif-1 recombinant protein was used as immunogen. | |
| 0.025 mg | |
| Apoptosis, Mitochondrial Mediated Pathway | |
| 51100 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction