missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10862-1-AP-20UL
This item is not returnable.
View return policy
Description
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
| Endoglin/CD105 | |
| Polyclonal | |
| Unconjugated | |
| ENG | |
| CD105, END, Endoglin, ENG, HHT1, ORW, ORW1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13805, 2022, 497010 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17813, Q63961 | |
| Eng | |
| Endoglin/CD105 Fusion Protein Ag1287 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction