missing translation for 'onlineSavingsMsg'
Learn More

Enalapril, Thermo Scientific™

Long-acting ACE inhibitor and antihypertensive

£85.80 - £303.00

Chemical Identifiers

CAS 75847-73-3
Molecular Formula C20H28N2O5
Molecular Weight (g/mol) 376.453
MDL Number MFCD00865774
InChI Key GBXSMTUPTTWBMN-XIRDDKMYSA-N
Synonym enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin
PubChem CID 5388962
ChEBI CHEBI:4784
IUPAC Name (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid
SMILES CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413469
SDS View Documents
Thermo Scientific Alfa Aesar
J60750.03
1g
£85.80
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15423469
SDS View Documents
Thermo Scientific Alfa Aesar
J60750.06
5g
£303.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Enalapril is used in the treatment of hypertension, diabetic nephropathy, and some types of chronic heart failure. It has been proven to protect the function of the kidneys in hypertension, heart failure, and diabetes, and may be used in the absence of hypertension for its kidney protective effects. It is widely used in chronic kidney failure.

Solubility
Soluble in water at 25mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Chemical Identifiers

75847-73-3
376.453
GBXSMTUPTTWBMN-XIRDDKMYSA-N
5388962
(2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid
C20H28N2O5
MFCD00865774
enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin
CHEBI:4784
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O

Specifications

75847-73-3
MFCD00865774
enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin
GBXSMTUPTTWBMN-XIRDDKMYSA-N
(2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid
5388962
376.45
C20H28N2O5
14,3567
Soluble in water at 25mg/ml
CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O
376.453
CHEBI:4784
Enalapril
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TW3665500

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.