Learn More
Enalapril, Thermo Scientific™
Long-acting ACE inhibitor and antihypertensive
Brand: Thermo Scientific Alfa Aesar J60750.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEnalapril is used in the treatment of hypertension, diabetic nephropathy, and some types of chronic heart failure. It has been proven to protect the function of the kidneys in hypertension, heart failure, and diabetes, and may be used in the absence of hypertension for its kidney protective effects. It is widely used in chronic kidney failure.
Solubility
Soluble in water at 25mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Chemical Identifiers
| 75847-73-3 | |
| 376.453 | |
| GBXSMTUPTTWBMN-XIRDDKMYSA-N | |
| 5388962 | |
| (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid |
| C20H28N2O5 | |
| MFCD00865774 | |
| enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin | |
| CHEBI:4784 | |
| CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O |
Specifications
| 75847-73-3 | |
| MFCD00865774 | |
| 14,3567 | |
| Soluble in water at 25mg/ml | |
| CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O | |
| 376.453 | |
| CHEBI:4784 | |
| Enalapril |
| C20H28N2O5 | |
| 5g | |
| enalapril, enalaprilum, vasotec, enalaprila, enalapril tn, unii-69pn84io1a, enalapril inn, enalapril richet, chembl578, enalaprilum inn-latin | |
| GBXSMTUPTTWBMN-XIRDDKMYSA-N | |
| (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid | |
| 5388962 | |
| 376.45 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.