missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EME1 Antibody (MTA31 7h2/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £307.00
Specifications
| Antigen | EME1 |
|---|---|
| Clone | MTA31 7h2/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684788
|
Novus Biologicals
NBP2-50386-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623409
|
Novus Biologicals
NBP2-50386 |
0.1 mL |
£307.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EME1 Monoclonal antibody specifically detects EME1 in Human samples. It is validated for Western BlotSpecifications
| EME1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| DNA Repair, Homologous Recombination | |
| PBS | |
| 146956 | |
| IgG1 | |
| Protein A purified |
| MTA31 7h2/1 | |
| Western Blot 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A | |
| Human EME1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title