missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EME1 Antibody (MTA31 7h2/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50386
This item is not returnable.
View return policy
Description
EME1 Monoclonal antibody specifically detects EME1 in Human samples. It is validated for Western Blot
Specifications
| EME1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| crossover junction endonuclease EME1, EC 3.1.22, EC 3.1.22.-, essential meiotic endonuclease 1 homolog 1 (S. pombe), essential meiotic endonuclease 1 homolog 2, FLJ31364, hMMS4, homolog of yeast EME1 endonuclease, MMS4, MMS4 homolog, MMS4L, SLX2 structure-specific endonuclease subunit homolog A, SLX2A | |
| Human EME1 | |
| 0.1 mL | |
| DNA Repair, Homologous Recombination | |
| 146956 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| MTA31 7h2/1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction