missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
Brand: Merck 14412
This item is not returnable.
View return policy
Description
Specifications
Adsorptive | |
100,000 NMWL | |
4.1 cm2 | |
10 Pack | |
0.98 in. |
Regenerated Cellulose | |
YM-100 | |
Protein purification, protein concentration, protein desalting | |
Ultrafiltration Disc | |
2.5 cm |