Learn More
Merck Adapter, male M Luer-Lok, nylon
Product Code. 10141731 Shop All Merck Products
Change view
Click to view available options
For Use With:
Millipore Filtration Systems
Unit Size:
Pack of 4
Product Code. For Use With unitSize
10141731 Millipore Filtration Systems Pack of 4
1 options
Product Code. 10141731
Brand: Merck XX1102503
Only null left
Add to basket
Add to basket
Male-to-male Luer-Lok™ adapter to convert a standard syringe into a hand vacuum pump
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Millipore Filtration Systems
Includes Adapter, male M Luer-Lok, nylon
Material Nylon
Sterility Non-sterile
Type Laboratory Filter Holders or Cyclones
Case Material Nylon